Merck
  • Fingolimod does not enhance cerebellar remyelination in the cuprizone model.
Fingolimod does not enhance cerebellar remyelination in the cuprizone model.
Journal of neuroimmunology (2015-07-23)
Maria Nordheim Alme, Agnes E Nystad, Lars Bø, Kjell-Morten Myhr, Christian A Vedeler, Stig Wergeland, Øivind Torkildsen
ABSTRACT
Fingolimod (FTY720) is approved for treatment of relapsing-remitting multiple sclerosis. In vitro studies have found that fingolimod stimulates remyelination in cerebellar slices, but in vivo animal studies have not detected any positive effect on cerebral remyelination. The discrepant findings could be a result of different mechanisms underlying cerebral and cerebellar remyelination. The cuprizone model for de- and remyelination was used to evaluate whether fingolimod had an impact on cerebellar remyelination in vivo. We found that fingolimod did not have any effect on cerebellar remyelination, number of mature oligodendrocytes, microglia or astrocytes when fed after cuprizone exposure.
Sigma-Aldrich
Anti-Neurofilament 70 kDa Antibody, clone DA2, culture supernatant, clone DA2, Chemicon®
Sigma-Aldrich
Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP) antibody produced in mouse, clone G-A-5, ascites fluid
Sigma-Aldrich
Anti-Neurite Outgrowth Inhibitor A Antibody, Chemicon®, from rabbit