Merck
  • Toward next generation plasma profiling via heat-induced epitope retrieval and array-based assays.
Toward next generation plasma profiling via heat-induced epitope retrieval and array-based assays.
Molecular & cellular proteomics : MCP (2010-08-05)
Jochen M Schwenk, Ulrika Igel, Maja Neiman, Hanno Langen, Charlotte Becker, Anders Bjartell, Fredrik Ponten, Fredrik Wiklund, Henrik Grönberg, Peter Nilsson, Mathias Uhlen
ABSTRACT
There is a need for high throughput methods for screening patient samples in the quest for potential biomarkers for diagnostics and patient care. Here, we used a combination of undirected target selection, antibody suspension bead arrays, and heat-induced epitope retrieval to allow for protein profiling of human plasma in a novel and systematic manner. Several antibodies were found to reveal altered protein profiles upon epitope retrieval at elevated temperatures with limits of detection improving into lower ng/ml ranges. In a study based on prostate cancer patients, several proteins with differential profiles were discovered and subsequently validated in an independent cohort. For one of the potential biomarkers, the human carnosine dipeptidase 1 protein (CNDP1), the differences were determined to be related to the glycosylation status of the targeted protein. The study shows a path of pursuit for large scale screening of biobank repositories in a flexible and proteome-wide fashion by utilizing heat-induced epitope retrieval and using an antibody suspension bead array format.