- Molecular docking and antiamnesic effects of nepitrin isolated from Rosmarinus officinalis on scopolamine-induced memory impairment in mice.
Molecular docking and antiamnesic effects of nepitrin isolated from Rosmarinus officinalis on scopolamine-induced memory impairment in mice.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2017-10-19)
Nasiara Karim, Imran Khan, Abeer Abdelhalim, Heba Abdel-Halim, Jane R Hanrahan
PMID29040957
ABSTRACT
Rosmarinus officinalis has long been known as the herb of remembrance. The present study was undertaken to investigate the anti-amnesic effects of nepitrin isolated from Rosmarinus officinalis using in-vivo models of Y-maze and novel object recognition test (NORT) along with in vitro antioxidant and acetylcholinesterase (AChE) and buterylcholinesterase (BuChE) inhibition potential. Nepitrin showed a concentration dependent inhibition of AChE and BuChE enzymes with IC