- miR-182 selectively targets HOXA10 in goat endometrial epithelium cells in vitro.
miR-182 selectively targets HOXA10 in goat endometrial epithelium cells in vitro.
Proper HOXA10 expression was essential for endometrial receptivity what was crucial for successful embryo implantation in mammalian. This study confirmed that miR-182 regulated the expression levels of HOXA10 by binding to its 3' UTR, selectively downregulated HOXA10 in goat endometrial epithelium cells (gEECs) but not stromal cell (gESCs) in vitro. However, HOXA10 and miR-182 both up-expressed in the goat endometrium at gestational day 15 (D15) compared with gestational day 5 (D5), suggesting that there were some other factors regulated the expression of HOXA10 during the development of goat endometrium in vivo. What's more, HOXA10 gene silencing (HOXA10-siRNA) resulted in gEECs apoptosis in vitro, and it regulated the protein levels of oestrogen receptor a (ERa), progesterone receptor B (PRb), insulin-like growth factor 1 receptor (IGF1R), BCL-2, pleiotrophin (PTN), AKT and p-JNK in gEECs. Furthermore, HOXA10 might regulate the protein levels of endometrial receptivity biomarker genes, including vascular endothelial growth factor (VEGF), osteopontin (OPN), cyclooxygenase-2 (COX-2) and prolactin receptor (PRLR) in gEECs. In conclusion, miR-182 targeted HOXA10 selectively in EECs in vitro, and HOXA10 played an important role in maintaining the function of EECs in dairy goats.