Merck
The Influence of Mineral Matrices on the Thermal Behavior of Glycine.
Origins of life and evolution of the biosphere : the journal of the International Society for the Study of the Origin of Life (2016-10-21)
Punam Dalai, Hannes Lukas Pleyer, Henry Strasdeit, Stefan Fox
ABSTRACT
On the Hadean-Early Archean Earth, the first islands must have provided hot and dry environments for abiotically formed organic molecules. The heat sources, mainly volcanism and meteorite impacts, were also available on Mars during the Noachian period. In recent work simulating this scenario, we have shown that neat glycine forms a black, sparingly water-soluble polymer ("thermomelanoid") when dry-heated at 200 °C under pure nitrogen. The present study explores whether relevant minerals and mineral mixtures can change this thermal behavior. Most experiments were conducted at 200 or 250 °C for 2 or 7 days. The mineral matrices used were phyllosilicates (Ca-montmorillonites SAz-1 and STx-1, Na-montmorillonite SAz-1-Na, nontronite NAu-1, kaolinite KGa-1), salts (NaCl, NaCl-KCl, CaCl
Sigma-Aldrich
Gly-Gly-Gly-Gly-Gly-Gly
Sigma-Aldrich
Gly-Gly-Gly-Gly