Merck
Immunological phenotype of the murine Lrba knockout.
Immunology and cell biology (2017-06-28)
Laura Gámez-Díaz, Julika Neumann, Fiona Jäger, Michele Proietti, Felicitas Felber, Pauline Soulas-Sprauel, Lisa Perruzza, Fabio Grassi, Tamara Kögl, Peter Aichele, Manfred Kilimann, Bodo Grimbacher, Sophie Jung
ABSTRACT
Biallelic mutations in the human lipopolysaccharide responsive beige-like anchor (LRBA) gene lead to a primary immunodeficiency known as LRBA deficiency, characterized by a broad range of clinical manifestations including autoimmunity, organomegaly, hypogammaglobulinemia and recurrent infections. Considering the phenotypic heterogeneity in patients and the severity of the disease, our aim was to assess the role of LRBA in immune cells and to understand the underlying pathomechanisms through the study of a Lrba knockout (Lrba
Sigma-Aldrich
Thyroglobulin from bovine thyroid, powder, ≥90% (agarose gel electrophoresis)