Merck
Effects of apolipoprotein M in uremic atherosclerosis.
Atherosclerosis (2017-09-04)
Markus Høybye Bosteen, Eva Martha Madsen Svarrer, Line Stattau Bisgaard, Torben Martinussen, Marie Madsen, Lars Bo Nielsen, Christina Christoffersen, Tanja Xenia Pedersen
ABSTRACT
Chronic kidney disease is characterized by uremia and causes premature death, partly due to accelerated atherosclerosis. Apolipoprotein (apo) M is a plasma carrier protein for the lipid sphingosine-1-phosphate (S1P). The Apom-S1P complex associates with HDL, and may contribute to its anti-atherosclerotic effects. The role of Apom/S1P in atherosclerosis is presently controversial and has not been explored in a uremic setting. We aimed to explore whether plasma concentrations of Apom/S1P are altered by uremia and whether Apom overexpression or deficiency affects classical and uremic atherosclerosis. Mild to moderate uremia was induced by subtotal nephrectomy (NX) in 86-92 Apoe-deficient mice that were either Apom-wild type, Apom-deficient, or overexpressed Apom (∼10 fold). The effects of uremia on plasma Apom/S1P and atherosclerosis were evaluated and compared to non-nephrectomized controls. Uremia increased plasma Apom by ∼25%, but not S1P. Plasma S1P was elevated by ∼300% in mice overexpressing Apom, and decreased by ∼25% in Apom-deficient mice. Apom overexpression augmented aortic root atherosclerosis and plasma cholesterol. In contrast, aortic arch atherosclerosis was unaffected by the Apom genotype. There was no effect of Apom-deficiency or Apom overexpression on uremic atherosclerosis. This study highlights the complexity of Apom/S1P in atherosclerosis and challenges the notion that the Apom/S1P complex is anti-atherogenic, at least in Apoe-deficient mice.
Sigma-Aldrich
Mayer′s Hematoxylin Solution