Merck
  • Effect of salt type and concentration on the growth and lipid content of Chlorella vulgaris in synthetic saline wastewater for biofuel production.
Effect of salt type and concentration on the growth and lipid content of Chlorella vulgaris in synthetic saline wastewater for biofuel production.
Bioresource technology (2017-06-27)
Jared Church, Jae-Hoon Hwang, Keug-Tae Kim, Rebecca McLean, You-Kwan Oh, Bora Nam, Jin Chul Joo, Woo Hyoung Lee
ABSTRACT
Microalgae can offer several benefits for wastewater treatment with their ability to produce large amounts of lipids for biofuel production and the high economic value of harvested biomass for biogas and fertilizer. This study found that salt concentration (∼45gL