Merck
Adrenocorticotropic hormone and 1,25-dihydroxyvitamin D
Laboratory investigation; a journal of technical methods and pathology (2017-07-25)
Irina L Tourkova, Li Liu, Nareerat Sutjarit, Quitterie C Larrouture, Jianhua Luo, Lisa J Robinson, Harry C Blair
ABSTRACT
To improve definition of the physical and hormonal support of bone formation, we studied differentiation of human osteoblasts in vitro at varying combinations of ACTH, 1α,25-dihydroxyvitamin D
Sigma-Aldrich
N,N′-Diphenylethylenediamine, 98%