Merck
  • Solubility and bioavailability enhancement of ciprofloxacin by induced oval-shaped mono-6-deoxy-6-aminoethylamino-β-cyclodextrin.
Solubility and bioavailability enhancement of ciprofloxacin by induced oval-shaped mono-6-deoxy-6-aminoethylamino-β-cyclodextrin.
Carbohydrate polymers (2017-03-08)
Jae Min Choi, Kyeonghui Park, Benel Lee, Daham Jeong, Someshwar D Dindulkar, Youngjin Choi, Eunae Cho, Seyeon Park, Jae-Hyuk Yu, Seunho Jung
ABSTRACT
Ciprofloxacin is a broad-spectrum fluoroquinolone antibiotic used to treat bacterial infections; however, its limited aqueous solubility inhibits its broader clinical uses. This study investigated the complexation effect of mono-6-deoxy-6-aminoethylamino-β-cyclodextrin on the aqueous solubility and bioavailability of ciprofloxacin. During complexation, the oval-shaped cavity induced by mono-aminoethylamine substitution on the primary rim of β-cyclodextrin, was considered to be a key factor according to NMR spectroscopy and molecular modeling studies. The ciprofloxacin with mono-6-deoxy-6-aminoethylamino-β-cyclodextrin complex was characterized using FE-SEM, DSC, FT-IR, T1 relaxation, 2D NOESY, and DOSY NMR spectroscopy and molecular modeling studies. The solubility property of ciprofloxacin complexed with mono-6-deoxy-6-aminoethylamino-β-cyclodextrin was enhanced by seven-fold compared to that of pure ciprofloxacin. Furthermore antibacterial activity of that complex against methicillin-resistant Staphylococcus aureus was enhanced and it clearly showed the growth inhibition. The mono-6-deoxy-6-aminoethylamino-β-cyclodextrin has the potential to be utilized for other oblong guest molecules besides ciprofloxacin based on the novel induced elliptical cavity.