Merck
  • Metabolism of T-2 Toxin in Farm Animals and Human In Vitro and in Chickens In Vivo Using Ultra High-Performance Liquid Chromatography- Quadrupole/Time-of-Flight Hybrid Mass Spectrometry Along with Online Hydrogen/Deuterium Exchange Technique.
Metabolism of T-2 Toxin in Farm Animals and Human In Vitro and in Chickens In Vivo Using Ultra High-Performance Liquid Chromatography- Quadrupole/Time-of-Flight Hybrid Mass Spectrometry Along with Online Hydrogen/Deuterium Exchange Technique.
Journal of agricultural and food chemistry (2017-07-25)
Shupeng Yang, Marthe De Boevre, Huiyan Zhang, Karl De Ruyck, Feifei Sun, Jinzhen Zhang, Yue Jin, Yanshen Li, Zhanhui Wang, Suxia Zhang, Jinhui Zhou, Yi Li, Sarah De Saeger
ABSTRACT
After being incubated with animal and human liver microsomes, metabolites of phase I and II were investigated. A comparison was performed by ultrahigh performance liquid chromatography-quadrupole/time-of-flight coupled to mass spectrometry (UHPLC-Q/TOF). Consequently, a total of four phase I metabolites and three glucuronide binding metabolites of T-2 toxin were discovered. Although a significant metabolic difference was observed among six species, HT-2 toxin was the major product in all species. In addition, the in vivo metabolism of T-2 toxin after oral administration was also investigated in chickens, In total, 18 metabolites were detected, of which 13 were novel, to our knowledge, and reported for the first time. To elucidate the structures of these metabolites, besides accurate mass data from their MS and MS
Sigma-Aldrich
Uridine 5′-diphosphoglucuronic acid ammonium salt, 98-100%