Merck
  • Synthesis, Spectral Characterization, DNA/ Protein Binding, DNA Cleavage, Cytotoxicity, Antioxidative and Molecular Docking Studies of Cu(II)Complexes Containing Schiff Base-bpy/Phen Ligands.
Synthesis, Spectral Characterization, DNA/ Protein Binding, DNA Cleavage, Cytotoxicity, Antioxidative and Molecular Docking Studies of Cu(II)Complexes Containing Schiff Base-bpy/Phen Ligands.
Journal of fluorescence (2017-01-18)
Berelli Anupama, Airva Aruna, Vijjulatha Manga, Sreekanth Sivan, Madamsetty Vijay Sagar, Ravula Chandrashekar
ABSTRACT
Ternary Cu(II) complexes [Cu(II)(L)(bpy)Cl] 1, [Cu(II)(L)(Phen)Cl] 2 [L = 2,3-dimethyl-1-phenyl-4(2 hydroxy-5-methyl benzylideneamino)-pyrazol-5-one, bpy = 2,2
Sigma-Aldrich
2-Hydroxy-5-methylbenzaldehyde, 98%