Merck
  • Silicon Supplementation Alters the Composition of Herbivore Induced Plant Volatiles and Enhances Attraction of Parasitoids to Infested Rice Plants.
Silicon Supplementation Alters the Composition of Herbivore Induced Plant Volatiles and Enhances Attraction of Parasitoids to Infested Rice Plants.
Frontiers in plant science (2017-08-05)
Jian Liu, Jiwei Zhu, Pengjun Zhang, Liwei Han, Olivia L Reynolds, Rensen Zeng, Jinhong Wu, Yue Shao, Minsheng You, Geoff M Gurr
ABSTRACT
Silicon (Si) is important in plant defenses that operate in a direct manner against herbivores, and work in rice (
Sigma-Aldrich
Nonyl acetate, ≥97%, FCC