Merck
  • 4-Amino-1,8-naphthalimide: a novel inhibitor of poly(ADP-ribose) polymerase and radiation sensitizer.
4-Amino-1,8-naphthalimide: a novel inhibitor of poly(ADP-ribose) polymerase and radiation sensitizer.
International journal of radiation biology (1999-02-11)
A Schlicker, P Peschke, A Bürkle, E W Hahn, J H Kim
ABSTRACT
Poly(ADP-ribose) polymerase (PARP; EC 2.4.2.30) is a chromatin-bound enzyme which is known to regulate chromatin structure by poly(ADP-ribosyl)ation of nuclear proteins, to facilitate DNA base excision repair, and to contribute to cellular recovery following DNA damage. Because inhibitors of PARP are able to potentiate the cell-killing effects of some DNA-damaging agents and to inhibit the repair of induced DNA strand breaks, such compounds may enhance the anti-tumour efficacy of radiotherapy or cytotoxic drug treatment. The PARP-inhibitory effects and radiosensitization of a new compound, 4-amino-1,8-naphthalimide (ANI), were examined. The inhibition of radiation-induced poly(ADP-ribosyl)ation (50 Gy; 60Co gamma-radiation) was evaluated by immunofluorescence assay using MoAb 10H directed against poly(ADP-ribose). Cell survival was assessed by colony forming assay (CFA) to determine the cytotoxicity of radiosensitization potential in exponentially growing hamster lung fibroblasts (V79), rat prostate carcinoma (R3327-AT1) and human prostate carcinoma (DU145) cells. At concentrations above 30 nmol x dm(-3) ANI, radiation-induced poly(ADP-ribose) was not detectable by immunofluorescence in V79, AT1 and DU145 cells. At the highest concentration tested for chronic exposure (20 micromol x dm(-3)), ANI was not cytotoxic and significantly potentiates the cytotoxicity of gamma-irradiation. The level of radiation enhancement was directly proportional to drug concentration. Survival curves for the three cell lines using 20 micromol x dm(-3) ANI revealed sensitizer enhancement ratios of 1.3 for V79, 1.5 for AT1 and 1.3 for DU145. In living cells, ANI is about 1000-fold more potent at inhibiting PARP activity compared with 3-aminobenzamide (3-ABA). CFA studies demonstrated that ANI is a radiation sensitizer at non-toxic and lower concentrations (20 micromol x dm(-3)) than 3-ABA (10 mmol x dm(-3)).
Sigma-Aldrich
4-Amino-1,8-naphthalimide