Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • A deleterious Nav1.1 mutation selectively impairs telencephalic inhibitory neurons derived from Dravet Syndrome patients.
A deleterious Nav1.1 mutation selectively impairs telencephalic inhibitory neurons derived from Dravet Syndrome patients.
eLife (2016-07-28)
Yishan Sun, Sergiu P Paşca, Thomas Portmann, Carleton Goold, Kathleen A Worringer, Wendy Guan, Karen C Chan, Hui Gai, Daniel Vogt, Ying-Jiun J Chen, Rong Mao, Karrie Chan, John Lr Rubenstein, Daniel V Madison, Joachim Hallmayer, Wendy M Froehlich-Santino, Jonathan A Bernstein, Ricardo E Dolmetsch
ABSTRACT
Dravet Syndrome is an intractable form of childhood epilepsy associated with deleterious mutations in SCN1A, the gene encoding neuronal sodium channel Nav1.1. Earlier studies using human induced pluripotent stem cells (iPSCs) have produced mixed results regarding the importance of Nav1.1 in human inhibitory versus excitatory neurons. We studied a Nav1.1 mutation (p.S1328P) identified in a pair of twins with Dravet Syndrome and generated iPSC-derived neurons from these patients. Characterization of the mutant channel revealed a decrease in current amplitude and hypersensitivity to steady-state inactivation. We then differentiated Dravet-Syndrome and control iPSCs into telencephalic excitatory neurons or medial ganglionic eminence (MGE)-like inhibitory neurons. Dravet inhibitory neurons showed deficits in sodium currents and action potential firing, which were rescued by a Nav1.1 transgene, whereas Dravet excitatory neurons were normal. Our study identifies biophysical impairments underlying a deleterious Nav1.1 mutation and supports the hypothesis that Dravet Syndrome arises from defective inhibitory neurons.
Sigma-Aldrich
Anti-Sodium Channel Antibody, Voltage Gated, Brain Type I, Chemicon®, from rabbit