Merck
  • Ultraviolet photochemistry of 2-bromothiophene explored using universal ionization detection and multi-mass velocity-map imaging with a PImMS2 sensor.
Ultraviolet photochemistry of 2-bromothiophene explored using universal ionization detection and multi-mass velocity-map imaging with a PImMS2 sensor.
The Journal of chemical physics (2017-07-10)
R A Ingle, C S Hansen, E Elsdon, M Bain, S J King, J W L Lee, M Brouard, C Vallance, R Turchetta, M N R Ashfold
ABSTRACT
The ultraviolet photochemistry of 2-bromothiophene (C
Sigma-Aldrich
2-Bromothiophene, 98%