Merck
  • Role of Cyclic Nucleotides in the Effect of Hydrogen Sulfide on Contractions of Rat Jejunum.
Role of Cyclic Nucleotides in the Effect of Hydrogen Sulfide on Contractions of Rat Jejunum.
Bulletin of experimental biology and medicine (2017-06-06)
D M Gabitova, I F Shaidullov, G I Sabirullina, M U Shafigullin, F G Sitdikov, G F Sitdikova
ABSTRACT
We studied the role of cyclic nucleotides in the influence of hydrogen sulfide (H
Sigma-Aldrich
1H-[1,2,4]Oxadiazolo[4,3-a]quinoxalin-1-one, powder
Sigma-Aldrich
8-(4-Chlorophenylthio)-guanosine 3′,5′-cyclic monophosphate sodium salt, ≥95% (HPLC), powder