Merck
  • New Method To Estimate Total Polyphenol Excretion: Comparison of Fast Blue BB versus Folin-Ciocalteu Performance in Urine.
New Method To Estimate Total Polyphenol Excretion: Comparison of Fast Blue BB versus Folin-Ciocalteu Performance in Urine.
Journal of agricultural and food chemistry (2017-05-06)
Daniel Hinojosa-Nogueira, Joaquín Muros, José A Rufián-Henares, Silvia Pastoriza
ABSTRACT
Polyphenols are bioactive substances of vegetal origin with a significant impact on human health. The assessment of polyphenol intake and excretion is therefore important. The Folin-Ciocalteu (F-C) method is the reference assay to measure polyphenols in foods as well as their excretion in urine. However, many substances can influence the method, making it necessary to conduct a prior cleanup using solid-phase extraction (SPE) cartridges. In this paper, we demonstrate the use of the Fast Blue BB reagent (FBBB) as a new tool to measure the excretion of polyphenols in urine. Contrary to F-C, FBBB showed no interference in urine, negating the time-consuming and costly SPE cleanup. In addition, it showed excellent linearity (r
Sigma-Aldrich
Enterodiol, ≥95.0% (HPLC)