Merck
  • Production of Biomass-Based Automotive Lubricants by Reductive Etherification.
Production of Biomass-Based Automotive Lubricants by Reductive Etherification.
ChemSusChem (2017-04-14)
Deepak Jadhav, Adam M Grippo, Sankaranarayanapillai Shylesh, Amit A Gokhale, John Redshaw, Alexis T Bell
ABSTRACT
Growing concern with the effects of CO
Sigma-Aldrich
2-Hexyl-1-decanol, 97%