Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Functional similarities between the dictyostelium protein AprA and the human protein dipeptidyl-peptidase IV.
Functional similarities between the dictyostelium protein AprA and the human protein dipeptidyl-peptidase IV.
Protein science : a publication of the Protein Society (2016-12-29)
Sarah E Herlihy, Yu Tang, Jonathan E Phillips, Richard H Gomer
ABSTRACT
Autocrine proliferation repressor protein A (AprA) is a protein secreted by Dictyostelium discoideum cells. Although there is very little sequence similarity between AprA and any human protein, AprA has a predicted structural similarity to the human protein dipeptidyl peptidase IV (DPPIV). AprA is a chemorepellent for Dictyostelium cells, and DPPIV is a chemorepellent for neutrophils. This led us to investigate if AprA and DPPIV have additional functional similarities. We find that like AprA, DPPIV is a chemorepellent for, and inhibits the proliferation of, D. discoideum cells, and that AprA binds some DPPIV binding partners such as fibronectin. Conversely, rAprA has DPPIV-like protease activity. These results indicate a functional similarity between two eukaryotic chemorepellent proteins with very little sequence similarity, and emphasize the usefulness of using a predicted protein structure to search a protein structure database, in addition to searching for proteins with similar sequences.
Sigma-Aldrich
FAP Active human, recombinant, expressed in baculovirus, ≥50% (SDS-PAGE)