Merck
Safety Evaluation of a Novel Strain of
Frontiers in microbiology (2017-04-04)
Ye Wang, Huimin Deng, Zhengchao Li, Yafang Tan, Yanping Han, Xiaoyi Wang, Zongmin Du, Yangyang Liu, Ruifu Yang, Yang Bai, Yujing Bi, Fachao Zhi
ABSTRACT
Commensal non-toxigenic
Sigma-Aldrich
4-Imidazoleacrylic acid, 99%
Sigma-Aldrich
Succinamic acid, 97%
Sigma-Aldrich
D-Glucosaminic acid, ≥98% (TLC)
Sigma-Aldrich
Maltose solution, BioReagent, ~20% in H2O, Molecular Biology
Sigma-Aldrich
β-Gentiobiose, ≥85% (remainder primarily α-anomer)