Merck
  • Apoptosis signal-regulating kinase 1 exhibits oncogenic activity in pancreatic cancer.
Apoptosis signal-regulating kinase 1 exhibits oncogenic activity in pancreatic cancer.
Oncotarget (2016-09-23)
Youguang Luo, Siqi Gao, Ziwei Hao, Yang Yang, Songbo Xie, Dengwen Li, Min Liu, Jun Zhou
ABSTRACT
Pancreatic cancer has an extremely grim prognosis, with an overall 5-year survival rate less than 5%, as a result of its rapid metastasis and late diagnosis. To combat this disease, it is crucial to better understand the molecular mechanisms that contribute to its pathogenesis. Herein, we report that apoptosis signal-regulating kinase 1 (ASK1) is overexpressed in pancreatic cancer tissues and that its expression correlates with the histological grade of pancreatic cancer. The expression of ASK1 is also elevated in pancreatic cancer cell lines at both protein and mRNA levels. In addition, ASK1 promotes the proliferation and stimulates the tumorigenic capacity of pancreatic cancer cells. These functions of ASK1 are abrogated by pharmacological inhibition of its kinase activity or by introduction of a kinase-dead mutation, suggesting that the kinase activity of ASK1 is required for its role in pancreatic cancer. However, the alteration of ASK1 expression or activity does not significantly affect the migration or invasion of pancreatic cancer cells. Collectively, these findings reveal a critical role for ASK1 in the development of pancreatic cancer and have important implications for the diagnosis and treatment of this malignancy.