Merck
  • IL-10 reduces apoptosis and extracellular matrix degradation after injurious compression of mature articular cartilage.
IL-10 reduces apoptosis and extracellular matrix degradation after injurious compression of mature articular cartilage.
Osteoarthritis and cartilage (2016-10-21)
P Behrendt, A Preusse-Prange, T Klüter, M Haake, B Rolauffs, A J Grodzinsky, S Lippross, B Kurz
ABSTRACT
The aim of this study was to examine whether anti-inflammatory interleukin-10 (IL-10) exerts chondroprotective effects in an in vitro model of a single mechanical injury of mature articular cartilage. Articular cartilage was harvested from the femoro-patellar groove of adult cows (Bos taurus) and cultured w/o bovine IL-10. After 24 h of equilibration explants were subjected to an axial unconfined compression (50% strain, velocity 2 mm/s, held for 10 s). After 96 h cell death was measured histomorphometrically (nuclear blebbing, NB) and the release of glycosaminoglycans (GAG, DMMB assay) and nitric oxide (NO, Griess-reagent) were analyzed. mRNA levels of matrix degrading enzymes and nitric oxide synthetase were measured by quantitative real time PCR. Differences between groups were calculated using a one-way ANOVA with a Bonferroni post hoc test. Injurious compression significantly increased the number of cells with NB, release of GAG and nitric oxide and expression of MMP-3, -13, ADAMTS-4 and NOS2. Administration of IL-10 significantly reduced the injury related cell death and release of GAG and NO, respectively. Expression of MMP-3, -13, ADAMTS-4 and NOS2 were significantly reduced. Joint injury is a complex process involving specific mechanical effects on cartilage as well as induction of an inflammatory environment. IL-10 prevented crucial mechanisms of chondrodegeneration induced by an injurious single compression. IL-10 might be a multipurpose drug candidate for the treatment of cartilage-related sports injuries or osteoarthritis (OA).
Sigma-Aldrich
1,9-Dimethyl-Methylene Blue zinc chloride double salt, Dye content 80 %