Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Impact of Gender in Renal Cell Carcinoma: The Relationship of FABP7 and BRN2 Expression with Overall Survival.
Impact of Gender in Renal Cell Carcinoma: The Relationship of FABP7 and BRN2 Expression with Overall Survival.
Clinical Medicine Insights. Oncology (2014-03-22)
Cheng Tan, Tatsuya Takayama, Naohisa Takaoka, Hiromi Fujita, Miki Miyazaki, Takayuki Sugiyama, Seiichiro Ozono
ABSTRACT
To investigate the relationship between gender differences in fatty acid-binding protein7 (FABP7) and BRN2 (POU class 3 homeobox 2) expression in renal cell carcinoma (RCC) and the prognosis of patients with RCC. immunohistochemical (IHC) staining as well as reverse transcription-polymerase chain reaction (RT-PCR) was performed in renal tissues from 103 patients (83 men, mean age = 63.6 years old; 20 women, mean age = 63.1 years old) underwent radical nephrectomy from January 1, 2001 through December 31, 2010. The probability of overall patient survival was estimated using the Kaplan-Meier method. FABP7 mRNA expression was more frequent in men (P = 0.07) while BRN2 protein expression was significantly more frequent in women (P = 0.029). In particular, FABP7 was expressed in 100% of G1 renal cell carcinoma both in mRNA and protein levels. In women, FABP7 (-) and BRN2 (+) groups had a worse prognosis both in mRNA level (P = 0.038) and protein level (P = 0.058). BRN2 was expressed 100% of papillary RCC both in mRNA and protein levels. Our results demonstrated that gender was a key factor in FABP7 and BRN2 expression in RCC, and the combination with FABP7 and BRN2 stratified by gender could be a new potential prognostic factor in patients with RCC.
Sigma-Aldrich
Anti-FABP7 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution