Merck
  • Regulation of heterochromatin transcription by Snail1/LOXL2 during epithelial-to-mesenchymal transition.
Regulation of heterochromatin transcription by Snail1/LOXL2 during epithelial-to-mesenchymal transition.
Molecular cell (2013-11-19)
Alba Millanes-Romero, Nicolás Herranz, Valentina Perrera, Ane Iturbide, Jordina Loubat-Casanovas, Jesús Gil, Thomas Jenuwein, Antonio García de Herreros, Sandra Peiró
ABSTRACT
Although heterochromatin is enriched with repressive traits, it is also actively transcribed, giving rise to large amounts of noncoding RNAs. Although these RNAs are responsible for the formation and maintenance of heterochromatin, little is known about how their transcription is regulated. Here, we show that the Snail1 transcription factor represses mouse pericentromeric transcription, acting through the H3K4 deaminase LOXL2. Since Snail1 plays a key role in the epithelial-to-mesenchymal transition (EMT), we analyzed the regulation of heterochromatin transcription in this process. At the onset of EMT, one of the major structural heterochromatin proteins, HP1α, is transiently released from heterochromatin foci in a Snail1/LOXL2-dependent manner, concomitantly with a downregulation of major satellite transcription. Moreover, preventing the downregulation of major satellite transcripts compromised the migratory and invasive behavior of mesenchymal cells. We propose that Snail1 regulates heterochromatin transcription through LOXL2, thus creating the favorable transcriptional state necessary for completing EMT.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
ChIPAb+ Trimethyl-Histone H3 (Lys9) - ChIP Validated Antibody and Primer Set, from rabbit