Merck
  • ASPM and CITK regulate spindle orientation by affecting the dynamics of astral microtubules.
ASPM and CITK regulate spindle orientation by affecting the dynamics of astral microtubules.
EMBO reports (2016-08-27)
Marta Gai, Federico T Bianchi, Cristiana Vagnoni, Fiammetta Vernì, Silvia Bonaccorsi, Selina Pasquero, Gaia E Berto, Francesco Sgrò, Alessandra Ma Chiotto, Laura Annaratone, Anna Sapino, Anna Bergo, Nicoletta Landsberger, Jacqueline Bond, Wieland B Huttner, Ferdinando Di Cunto
ABSTRACT
Correct orientation of cell division is considered an important factor for the achievement of normal brain size, as mutations in genes that affect this process are among the leading causes of microcephaly. Abnormal spindle orientation is associated with reduction of the neuronal progenitor symmetric divisions, premature cell cycle exit, and reduced neurogenesis. This mechanism has been involved in microcephaly resulting from mutation of ASPM, the most frequently affected gene in autosomal recessive human primary microcephaly (MCPH), but it is presently unknown how ASPM regulates spindle orientation. In this report, we show that ASPM may control spindle positioning by interacting with citron kinase (CITK), a protein whose loss is also responsible for severe microcephaly in mammals. We show that the absence of CITK leads to abnormal spindle orientation in mammals and insects. In mouse cortical development, this phenotype correlates with increased production of basal progenitors. ASPM is required to recruit CITK at the spindle, and CITK overexpression rescues ASPM phenotype. ASPM and CITK affect the organization of astral microtubules (MT), and low doses of MT-stabilizing drug revert the spindle orientation phenotype produced by their knockdown. Finally, CITK regulates both astral-MT nucleation and stability. Our results provide a functional link between two established microcephaly proteins.
Sigma-Aldrich
Anti-Tbr2 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-γ-Tubulin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone B-5-1-2, ascites fluid