Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Synthetic Lethal Screen Demonstrates That a JAK2 Inhibitor Suppresses a BCL6-dependent IL10RA/JAK2/STAT3 Pathway in High Grade B-cell Lymphoma.
Synthetic Lethal Screen Demonstrates That a JAK2 Inhibitor Suppresses a BCL6-dependent IL10RA/JAK2/STAT3 Pathway in High Grade B-cell Lymphoma.
The Journal of biological chemistry (2016-06-09)
Daniel Beck, Jenny Zobel, Ruth Barber, Sian Evans, Larissa Lezina, Rebecca L Allchin, Matthew Blades, Richard Elliott, Christopher J Lord, Alan Ashworth, Andrew C G Porter, Simon D Wagner
ABSTRACT
We demonstrate the usefulness of synthetic lethal screening of a conditionally BCL6-deficient Burkitt lymphoma cell line, DG75-AB7, with a library of small molecules to determine survival pathways suppressed by BCL6 and suggest mechanism-based treatments for lymphoma. Lestaurtinib, a JAK2 inhibitor and one of the hits from the screen, repressed survival of BCL6-deficient cells in vitro and reduced growth and proliferation of xenografts in vivo BCL6 deficiency in DG75-AB7 induced JAK2 mRNA and protein expression and STAT3 phosphorylation. Surface IL10RA was elevated by BCL6 deficiency, and blockade of IL10RA repressed STAT3 phosphorylation. Therefore, we define an IL10RA/JAK2/STAT3 pathway each component of which is repressed by BCL6. We also show for the first time that JAK2 is a direct BCL6 target gene; BCL6 bound to the JAK2 promoter in vitro and was enriched by ChIP-seq. The place of JAK2 inhibitors in the treatment of diffuse large B-cell lymphoma has not been defined; we suggest that JAK2 inhibitors might be most effective in poor prognosis ABC-DLBCL, which shows higher levels of IL10RA, JAK2, and STAT3 but lower levels of BCL6 than GC-DLBCL and might be usefully combined with novel approaches such as inhibition of IL10RA.
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Benzyl alcohol, ACS reagent, ≥99.0%