Merck
  • Mutations of the SCO1 gene in mitochondrial cytochrome c oxidase deficiency with neonatal-onset hepatic failure and encephalopathy.
Mutations of the SCO1 gene in mitochondrial cytochrome c oxidase deficiency with neonatal-onset hepatic failure and encephalopathy.
American journal of human genetics (2000-10-03)
I Valnot, S Osmond, N Gigarel, B Mehaye, J Amiel, V Cormier-Daire, A Munnich, J P Bonnefont, P Rustin, A Rötig
ABSTRACT
Cytochrome c oxidase (COX) catalyzes both electron transfer from cytochrome c to molecular oxygen and the concomitant vectorial proton pumping across the inner mitochondrial membrane. Studying a large family with multiple cases of neonatal ketoacidotic comas and isolated COX deficiency, we have mapped the disease locus to chromosome 17p13.1, in a region encompassing two candidate genes involved in COX assembly-namely, SCO1 and COX10. Mutation screening revealed compound heterozygosity for SCO1 gene mutations in the patients. The mutated allele, inherited from the father, harbored a 2-bp frameshift deletion (DeltaGA; nt 363-364) resulting in both a premature stop codon and a highly unstable mRNA. The maternally inherited mutation (C520T) changed a highly conserved proline into a leucine in the protein (P174L). This proline, adjacent to the CxxxC copper-binding domain of SCO1, is likely to play a crucial role in the tridimentional structure of the domain. Interestingly, the clinical presentation of SCO1-deficient patients markedly differs from that of patients harboring mutations in other COX assembly and/or maturation genes.