Merck
  • Functional genetic screens for enhancer elements in the human genome using CRISPR-Cas9.
Functional genetic screens for enhancer elements in the human genome using CRISPR-Cas9.
Nature biotechnology (2016-01-12)
Gozde Korkmaz, Rui Lopes, Alejandro P Ugalde, Ekaterina Nevedomskaya, Ruiqi Han, Ksenia Myacheva, Wilbert Zwart, Ran Elkon, Reuven Agami
ABSTRACT
Systematic identification of noncoding regulatory elements has, to date, mainly relied on large-scale reporter assays that do not reproduce endogenous conditions. We present two distinct CRISPR-Cas9 genetic screens to identify and characterize functional enhancers in their native context. Our strategy is to target Cas9 to transcription factor binding sites in enhancer regions. We identified several functional enhancer elements and characterized the role of two of them in mediating p53 (TP53) and ERα (ESR1) gene regulation. Moreover, we show that a genomic CRISPR-Cas9 tiling screen can precisely map functional domains within enhancer elements. Our approach expands the utility of CRISPR-Cas9 to elucidate the functions of the noncoding genome.
Sigma-Aldrich
Nutlin-3a, ≥98% (HPLC)
Sigma-Aldrich
CRISPR/Cas9 Products and Services, Design and order CRISPR gRNA, Cas9, screening libraries, controls and companion products. Formats include plant, lentivirus, IVT-RNA, plasmid, synthetic, and protein.