Merck
  • Application of the Rh(II) cyclization/cycloaddition cascade for the total synthesis of (+/-)-aspidophytine.
Application of the Rh(II) cyclization/cycloaddition cascade for the total synthesis of (+/-)-aspidophytine.
Organic letters (2006-07-14)
José M Mejía-Oneto, Albert Padwa
ABSTRACT
[Structure: see text] A new strategy for the synthesis of (+/-)-aspidophytine has been developed and is based on a Rh(II)-catalyzed cyclization/dipolar cycloaddition sequence. The resulting [3+2]-cycloadduct undergoes an efficient Lewis acid mediated cascade that rapidly provides the complete skeleton of aspidophytine. The synthesis also features a mild decarbomethoxylation reaction.
Sigma-Aldrich
N-Phenyl-bis(trifluoromethanesulfonimide), 99%