Merck
  • Therapeutic preparations of IVIg contain naturally occurring anti-HLA-E antibodies that react with HLA-Ia (HLA-A/-B/-Cw) alleles.
Therapeutic preparations of IVIg contain naturally occurring anti-HLA-E antibodies that react with HLA-Ia (HLA-A/-B/-Cw) alleles.
Blood (2013-01-12)
Mepur H Ravindranath, Paul I Terasaki, Tho Pham, Vadim Jucaud, Satoru Kawakita
ABSTRACT
The US Food and Drug Administration approved intravenous immunoglobulin (IVIg), extracted from the plasma of thousands of blood donors, for removing HLA antibodies (Abs) in highly sensitized patients awaiting organ transplants. Since the blood of healthy individuals has HLA Abs, we tested different IVIg preparations for reactivity to HLA single antigen Luminex beads. All preparations showed high levels of HLA-Ia and -Ib reactivity. Since normal nonalloimmunized males have natural antibodies to the heavy chains (HCs) of HLA antigens, the preparations were then tested against iBeads coated only with intact HLA antigens. All IVIg preparations varied in level of antibody reactivity to intact HLA antigens. We raised monoclonal Abs against HLA-E that mimicked IVIg's HLA-Ia and HLA-Ib reactivity but reacted only to HLA-I HCs. Inhibition experiments with synthetic peptides showed that HLA-E shares epitopes with HLA-Ia alleles. Importantly, depleting anti-HLA-E Abs from IVIg totally eliminated the HLA-Ia reactivity of IVIg. Since anti-HLA-E mAbs react with HLA-Ia, they might be useful in suppressing HLA antibody production, similar to the way anti-RhD Abs suppress production. At the same time, anti-HLA-E mAb, which reacts only to HLA-I HCs, is unlikely to produce transfusion-related acute lung injury, in contrast to antibodies reacting to intact-HLA.