Merck
  • A practical method for the synthesis of highly enantioenriched trans-1,2-amino alcohols.
A practical method for the synthesis of highly enantioenriched trans-1,2-amino alcohols.
Organic letters (2013-06-08)
James A Birrell, Eric N Jacobsen
ABSTRACT
A highly enantioselective addition of phenyl carbamate to meso-epoxides has been developed to efficiently generate protected trans-1,2-amino alcohols. This transformation is promoted by an oligomeric (salen)Co-OTf catalyst and has been used to prepare two useful 2-aminocycloalkanol hydrochlorides in enantiopure form on a multigram scale from commercially available starting materials.