Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Measurement of Postreplicative DNA Metabolism and Damage in the Rodent Brain.
Measurement of Postreplicative DNA Metabolism and Damage in the Rodent Brain.
Chemical research in toxicology (2015-10-09)
Jay P Patel, Mark L Sowers, Jason L Herring, Jacob A Theruvathu, Mark R Emmett, Bridget E Hawkins, Kangling Zhang, Douglas S DeWitt, Donald S Prough, Lawrence C Sowers
ABSTRACT
The DNA of all organisms is metabolically active due to persistent endogenous DNA damage, repair, and enzyme-mediated base modification pathways important for epigenetic reprogramming and antibody diversity. The free bases released from DNA either spontaneously or by base excision repair pathways constitute DNA metabolites in living tissues. In this study, we have synthesized and characterized the stable-isotope standards for a series of pyrimidines derived from the normal DNA bases by oxidation and deamination. We have used these standards to measure free bases in small molecule extracts from rat brain. Free bases are observed in extracts, consistent with both endogenous DNA damage and 5-methylcytosine demethylation pathways. The most abundant free base observed is uracil, and the potential sources of uracil are discussed. The free bases measured in tissue extracts constitute the end product of DNA metabolism and could be used to reveal metabolic disturbances in human disease.
Sigma-Aldrich
Potassium perruthenate
Sigma-Aldrich
Orotic acid, ≥98% (titration), anhydrous