Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Iridium(III) silyl alkyl and iridacyclic compounds supported by 4,4'-di-tert-butyl-2,2'-bipyridyl.
Iridium(III) silyl alkyl and iridacyclic compounds supported by 4,4'-di-tert-butyl-2,2'-bipyridyl.
Chemistry (Weinheim an der Bergstrasse, Germany) (2006-09-23)
Yiu-Keung Sau, Hung-Kay Lee, Ian D Williams, Wa-Hung Leung
ABSTRACT
Treatment of IrCl(3)x H(2)O with one equivalent of 4,4'-di-tert-butyl-2,2'-bipyridyl (dtbpy) in N,N-dimethylformamide (dmf) afforded [IrCl(3)(dmf)(dtbpy)] (1). Alkylation of 1 with Me(3)SiCH(2)MgCl resulted in C--Si cleavage of the Me(3)SiCH(2) group and formation of the Ir(III) silyl dialkyl compound [Ir(CH(2)SiMe(3))(dtbpy)(Me)(SiMe(3))] (2), which reacted with tBuNC to afford [Ir(tBuNC)(CH(2)SiMe(3))(dtbpy)(Me)(SiMe(3))] ([2(tBuNC)]). Reaction of 2 with phenylacetylene afforded dimeric [{Ir(C[triple chemical bond]CPh)(dtbpy)(SiMe(3))}(2)(mu-C[triple chemical bond]CPh)(2)] (3), in which the bridging PhC[triple chemical bond]C(-) ligands are bound to Ir in a mu-sigma:pi fashion. Alkylation of 1 with PhMe(2)CCH(2)MgCl afforded the cyclometalated compound [Ir(dtbpy)(CH(2)CMe(2)C(6)H(4))(2-C(6)H(4)CMe(3))] (4), which features an agostic interaction between the Ir center and the 2-tert-butylphenyl ligand. The cyclic voltammogram of 4 in CH(2)Cl(2) shows a reversible Ir(IV)-Ir(III) couple at about 0.02 V versus ferrocenium/ferrocene. Oxidation of 4 in CH(2)Cl(2) with silver triflate afforded an Ir(IV) species that exhibits an anisotropic electron paramagnetic resonance (EPR) signal in CH(2)Cl(2) glass at 4 K with g( parallel)=2.430 and g( perpendicular)=2.110. Protonation of 4 with HCl and p-toluenesulfonic acid (HOTs) afforded [{Ir(dtbpy)(CH(2)CMe(2)Ph)Cl}(2)(mu-Cl)(2)] (5) and [Ir(dtbpy)(CH(2)CMe(2)Ph)(OTs)(2)] (6), respectively. Reaction of 5 with Li[BEt(3)H] gave the cyclometalated complex [{Ir(dtbpy)(CH(2)CMe(2)C(6)H(4))}(2)(mu-Cl)(2)] (7). Reaction of 4 with tetracyanoethylene in refluxing toluene resulted in electrophilic substitution of the iridacycle by C(2)(CN)(3) with formation of [Ir(dtbpy)(CH(2)CMe(2)C(6)H(3){4-C(2)(CN)(3)})(2-C(6)H(4)CMe(3))] (8). Reaction of 4 with diethyl maleate in refluxing toluene gave the iridafuran compound [Ir(dtbpy)(CH(2)CMe(2)C(6)H(4)){kappa(2)(C,O)-C(CO(2)Et)CH(CO(2)Et)}] (9). Treatment of 9 with 2,6-dimethylphenyl isocyanide (xylNC) led to cleavage of the iridafuran ring and formation of [Ir(dtbpy)(CH(2)CMe(2)C(6)H(4)){C(CO(2)Et)CH(CO(2)Et)}(xylNC)] (10). Protonation of 9 with HBF(4) afforded the dinuclear neophyl complex [(Ir(dtbpy)(CH(2)CMe(2)Ph){kappa(2)(C,O)-C(CO(2)Et)CH(CO(2)Et)})(2)][BF(4)](2) (11). The solid-state structures of complexes 2-5 and 8-11 have been determined.
Sigma-Aldrich
4,4′-Di-tert-butyl-2,2′-dipyridyl, 98%