Merck
Get up to 22% off for Pi Day until 3/26.Save Now
Rictor is an independent prognostic factor for endometrial carcinoma.
International journal of clinical and experimental pathology (2014-06-27)
Shan-Yun Wen, Chang-Hua Li, Yan-Li Zhang, Yu-Hai Bian, Li Ma, Qiu-Lin Ge, Yin-Cheng Teng, Zhi-Gang Zhang
ABSTRACT
Early-stage endometrial carcinoma (EC) patients have a high cure rate; however, those with high-risk factors may have poor prognosis. Thus, there is an urgent need for searching for new prognostic molecules to more accurately predict survival of patients. We detected the Rictor mRNA expression level in 30 fresh EC tissue and 17 normal endometrial tissue samples with real-time quantitative RT-PCR and Rictor protein expression level in 134 (test cohort) and 115 (validation cohort) paraffin tissue samples by immunohistochemistry, analyzed the correlation between variables and overall survival (OS) using Cox proportional hazards regression, compared the prognostic accuracy of Rictor with other clinicopathological risk factors by logistic regression. The results showed that Rictor mRNA expression of EC is higher than that of normal endometrium; Rictor protein expression level was closely correlated with FIGO stage, grade and vascular invasion in both cohorts; a univariate analysis showed that the pathological type, stage, grade, vascular invasion, lymphatic metastasis and Rictor were predictors of OS in both cohorts; furthermore, multivariate Cox proportional hazards regression analysis indicated that vascular invasion and Rictor were independent prognostic factors for EC in both cohorts; an ROX curve comparison showed that the area under the curve (AUC) for Rictor combined with other clinicopathological prognostic factors was higher than any individual factor or other clinicopathological prognostic factors' combination. Based on the above data, we concluded that Rictor is an independent prognostic factor for EC. It combined with other clinicopathological risk factors was a stronger prognostic model than individual risk factor or their combination.