Merck
  • Toll-like receptor 3 regulates NK cell responses to cytokines and controls experimental metastasis.
Toll-like receptor 3 regulates NK cell responses to cytokines and controls experimental metastasis.
Oncoimmunology (2015-09-26)
Camille Guillerey, Melvyn T Chow, Kim Miles, Stuart Olver, Jaclyn Sceneay, Kazuyoshi Takeda, Andreas Möller, Mark J Smyth
ABSTRACT
The Toll-like receptor 3 (TLR3) agonist poly(I:C) is a promising adjuvant for cancer vaccines due to its induction of potent antitumor responses occurring primarily through the activation of dendritic cells (DCs) and natural killer (NK) cells. However, little is known about the role of TLR3 sensing of endogenous ligands in innate tumor immunosurveillance. Here, we investigated whether TLR3 could modulate immune responses and facilitate tumor control without administration of an agonist. We observed only limited impact of TLR3 deficiency on spontaneous carcinogenesis and primary growth of B16F10, E0771 or MC38 tumors when injected subcutaneously to mice. Nevertheless, TLR3 was observed to limit experimental B16F10 lung metastasis, an immunologic constraint dependent on both IFNγ secretion and NK cells. Interestingly, we observed that NK cells derived from
Sigma-Aldrich
6-Maleimidohexanoic acid, 90% (GC)
Sigma-Aldrich
Interleukin-18 from rat, >97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder