Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Risk factors, clinical features and outcomes of visceral leishmaniasis in solid-organ transplant recipients: a retrospective multicenter case-control study.
Risk factors, clinical features and outcomes of visceral leishmaniasis in solid-organ transplant recipients: a retrospective multicenter case-control study.
Clinical microbiology and infection : the official publication of the European Society of Clinical Microbiology and Infectious Diseases (2015-02-01)
W Clemente, E Vidal, E Girão, A S D Ramos, F Govedic, E Merino, P Muñoz, N Sabé, C Cervera, G F Cota, E Cordero, A Mena, M Montejo, F López-Medrano, J M Aguado, P Fernandes, M Valerio, J Carratalá, A Moreno, J Oliveira, P H O Mourão, J Torre-Cisneros
ABSTRACT
Visceral leishmaniasis (VL) is a rare disease in solid-organ transplant (SOT) recipients. Therefore, little is known about the risk factors and disease behavior in the transplant setting. This multicenter, matched case-control study (1:2 ratio) was designed to determine the risk factors, clinical features and outcomes of VL among this population. Control and case subjects were matched by center, transplant type and timing. Thirty-six VL cases were identified among 25 139 SOT recipients (0.1%). VL occurred 5.7-fold more frequently in Brazil than in Spain, presenting a median time of 11 months after transplantation. High-dose prednisone in the preceding 6 months was associated with VL. Patients were diagnosed over 1 month after symptom onset in 25% of cases. Thirty-one patients (86%) were febrile upon diagnosis, 81% exhibited visceromegaly and 47% showed pancytopenia. Concomitant infection was common. Parasites were identified in 89% of patients; the remaining patients were diagnosed by serology. The majority of the patients received amphotericin B. Relapses occurred in 25.7% of cases, and the crude mortality rate was 2.8%. VL after SOT is related to the VL prevalence in the general population. Delayed diagnosis frequently occurs. Liposomal amphotericin is the most commonly used therapy; mortality is low, although relapses are common.
Sigma-Aldrich
Vinyl laurate, ≥99.0% (GC)
Sigma-Aldrich
Vinyl laurate, Wacker Chemie AG, ≥98% (GC)