Merck
  • Palladium-catalyzed carbonylations of aryl bromides using paraformaldehyde: synthesis of aldehydes and esters.
Palladium-catalyzed carbonylations of aryl bromides using paraformaldehyde: synthesis of aldehydes and esters.
Angewandte Chemie (International ed. in English) (2014-07-30)
Kishore Natte, Andreas Dumrath, Helfried Neumann, Matthias Beller
ABSTRACT
Carbonylation reactions represent useful tools for organic synthesis. However, the necessity to use gaseous carbon monoxide is a disadvantage for most organic chemists. To solve this problem, novel protocols have been developed for conducting palladium-catalyzed reductive carbonylations of aryl bromides and alkoxycarbonylations using paraformaldehyde as an external CO source (CO gas free). Hence, aromatic aldehydes and esters were synthesized in moderate to good yields.
Sigma-Aldrich
Paraformaldehyde, powder, 95%