Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • A genome-wide association study of total serum and mite-specific IgEs in asthma patients.
A genome-wide association study of total serum and mite-specific IgEs in asthma patients.
PloS one (2013-08-24)
Jeong-Hyun Kim, Hyun Sub Cheong, Jong Sook Park, An-Soo Jang, Soo-Taek Uh, Yong-Hoon Kim, Mi-Kyeong Kim, Inseon S Choi, Sang Heon Cho, Byoung Whui Choi, Joon Seol Bae, Choon-Sik Park, Hyoung Doo Shin
ABSTRACT
Immunoglobulin E (IgE) is one of the central players in asthma and allergic diseases. Although the serum IgE level, a useful endophenotype, is generally increased in patients with asthma, genetic factors influencing IgE regulation in asthma are still not fully understood. To identify the genetic variations associated with total serum and mite-specific IgEs in asthmatics, a genome-wide association study (GWAS) of 657,366 single nucleotide polymorphisms (SNPs) was performed in 877 Korean asthmatics. This study found that several new genes might be associated with total IgE in asthmatics, such as CRIM1 (rs848512, P = 1.18×10(-6); rs711254, P = 6.73×10(-6)), ZNF71 (rs10404342, P = 7.60×10(-6)), TLN1 (rs4879926, P = 7.74×10(-6)), and SYNPO2 (rs1472066, P = 8.36×10(-6); rs1038770, P = 8.66×10(-6)). Regarding the association of specific IgE to house dust mites, it was observed that intergenic SNPs nearby to OPRK1 and LOC730217 might be associated with Dermatophagoides pteronyssinus (D.p.) and Dermatophagoides farinae (D.f.) in asthmatics, respectively. In further pathway analysis, the phosphatidylinositol signaling system and adherens junction pathways were estimated to play a role in the regulation of total IgE levels in asthma. Although functional evaluations and replications of these results in other populations are needed, this GWAS of serum IgE in asthmatics could facilitate improved understanding of the role of the newly identified genetic variants in asthma and its related phenotypes.