Merck
  • PD-1 deletion restores susceptibility to experimental autoimmune encephalomyelitis in miR-155-deficient mice.
PD-1 deletion restores susceptibility to experimental autoimmune encephalomyelitis in miR-155-deficient mice.
International immunology (2014-03-22)
Jinyu Zhang, Michel Y Braun
ABSTRACT
MiR-155 (-/-) mice are highly resistant to experimental autoimmune encephalomyelitis (EAE), while Pdcd1 (-/-) mice develop a more severe form of the disease. To determine the conflicting roles of these two molecules in the disease, we generated miR-155 (-/-) Pdcd1 (-/-) double knockout (DKO) mice. We found that ablation of programmed cell death protein 1 (PD-1) expression in miR-155-deficient mice restored the susceptibility to EAE. The increased severity of the disease in DKO mice was accompanied by an enhanced T-cell infiltration into the brain as well as an increased production of pro-inflammatory cytokines IFN-γ and IL-17. Furthermore, the major contribution of the DKO to EAE was T-cell intrinsic since adoptive transfer of CD4(+) T cells from DKO donors promoted the disease in lymphopenic recipients. These results define PD-1 deficiency in miR-155 (-/-) mice as a promoting factor of autoimmune inflammation by increasing antigen-driven T-cell expansion and infiltration.
Sigma-Aldrich
Human IL-17 ELISA Kit, for serum, plasma, cell culture supernatant and urine