Merck
  • The anti-ictogenic effects of levetiracetam are mirrored by interictal spiking and high-frequency oscillation changes in a model of temporal lobe epilepsy.
The anti-ictogenic effects of levetiracetam are mirrored by interictal spiking and high-frequency oscillation changes in a model of temporal lobe epilepsy.
Seizure (2015-02-04)
Maxime Lévesque, Charles Behr, Massimo Avoli
ABSTRACT
Mesial temporal lobe epilepsy (MTLE) is the most prevalent type of partial epileptic disorders. In this study, we have analyzed the impact of levetiracetam (LEV) in the pilocarpine model of MTLE. Sprague-Dawley rats (n=19) were injected with pilocarpine (380 mg/kg, i.p.) to induce a status epilepticus. Twelve animals were used as controls and seven were treated with LEV. They were implanted with bipolar electrodes in the CA3 subfield of the hippocampus, entorhinal cortex (EC), dentate gyrus (DG) and subiculum and EEG-video monitored continuously from day 4 to day 14 after SE. Only 29% of LEV-treated animals had seizures compared to all controls following a latent period that was similar in duration. Seizure rates were lower in LEV-treated animals. In LEV-treated animals without seizures, lower interictal spike rates were found in all regions compared to controls. Analysis of interictal high-frequency oscillations (HFO s) revealed that LEV-treated animals without seizures had lower rates of interictal spikes with ripples (80-200 Hz) in CA3, EC and subiculum (p<0.01), whereas rates of interictal spikes with fast ripples (250-500 Hz) were significantly lower in CA3 and subiculum, compared to controls. Our findings indicate that the anti-ictogenic properties of LEV are mirrored by decreases of interictal spike rate in temporal lobe regions, and are accompanied by subregion-specific decreases of HFO occurrence in CA3 and subiculum. Overall, this evidence suggest that LEV may inhibit neural network activity in regions that are known to play important roles in MTLE.
Product Description
Pilocarpine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Pilocarpine hydrochloride, ≥99% (titration), powder