Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • The effects of an epithelial barrier protective cationic aerosol on allergen-induced airway inflammation in asthma: a randomized, placebo-controlled clinical trial.
The effects of an epithelial barrier protective cationic aerosol on allergen-induced airway inflammation in asthma: a randomized, placebo-controlled clinical trial.
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology (2014-08-19)
P Nair, S Denis, L Cancelliere, K Radford, A Efthimiadis, M Rosano, J Hanrahan
ABSTRACT
Inhaled cationic airway lining modulator (iCALM) is a cationic aerosol therapy comprised of 1.29% calcium chloride dissolved in 0.9% isotonic saline that enhances the biophysical barrier function of the airway lining fluid and primes the host defense response. It's ability to attenuate bronchitis caused by inhaled particles was investigated using an allergen-inhalation model in a proof-of-concept study. In a randomized, double-blind, placebo-controlled cross-over trial of 6 mild atopic steroid-naïve asthmatic subjects, 3 doses of iCALM were well tolerated and they attenuated allergen-induced increase in sputum eosinophils, and levels of IL-5, MCP-1 and eotaxin. This study provides an opportunity to investigate the role of enhancing epithelial barrier to decrease airway inflammation provoked by inhaled particles in a variety of airway diseases.
Sigma-Aldrich
Mouse Eotaxin ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Human Eotaxin / CCL11 ELISA Kit, for serum, plasma, cell culture supernatant and urine