Merck
  • Unexpected inverse correlation between Native American ancestry and Asian American variants of HPV16 in admixed Colombian cervical cancer cases.
Unexpected inverse correlation between Native American ancestry and Asian American variants of HPV16 in admixed Colombian cervical cancer cases.
Infection, genetics and evolution : journal of molecular epidemiology and evolutionary genetics in infectious diseases (2014-12-03)
Esteban A Lopera, Armando Baena, Victor Florez, Jehidys Montiel, Constanza Duque, Tatiana Ramirez, Mauricio Borrero, Carlos M Cordoba, Fredy Rojas, Rene Pareja, Astrid M Bedoya, Gabriel Bedoya, Gloria I Sanchez
ABSTRACT
European (E) variants of HPV 16 are evenly distributed among world regions, meanwhile Non-European variants such as European-Asian (EAs), Asian American (AA) and African (Af) are mostly confined to Eastern Asia, The Americas and African regions respectively. Several studies have shown that genetic variation of HPV 16 is associated with the risk of cervical cancer, which also seems to be dependent on the population. This relationship between ethnicity and variants have led to the suggestion that there is co-evolution of variants with humankind. Our aim was to evaluate the relationship between the individual ancestry proportion and infection with HPV 16 variants in cervical cancer. We examined the association between ancestry and HPV 16 variants in samples of 82 cervical cancer cases from different regions of Colombia. Individual ancestry proportions (European, African and Native American) were estimated by genotyping 106 ancestry informative markers. Variants were identified by PCR amplification of the E6 gene, followed by reverse line blot hybridization (RLB) with variants specific probes. Overall European (E) and Asian American (AA) variants frequency was 66.5% and 33.5% respectively. Similar distribution was observed in cases with higher proportions of European or African ancestry. A higher Native American ancestry was significantly associated with higher frequency of E variants (median ancestry>23.6%, Age and place of birth adjusted OR: 3.55, 95% CI: 1.26-10.03, p=0.01). Even further, an inverse geographic correlation between Native American ancestry and frequency of infections with AA variants was observed (ρ=-0.825, p=0.008). Regions with higher proportion of Native American ancestry had a lower frequency of AA variants of HPV 16. This study suggests replacement of AA variants by E variants of human papillomavirus 16 in cervical cancer cases with high Native American ancestry.
Sigma-Aldrich
A1120, ≥98% (HPLC), powder