Merck
CXCL3 is a potential target for breast cancer metastasis.
Current cancer drug targets (2014-03-13)
Amanda Lay Pin See, Poh Kuan Chong, Ssu-Yi Lu, Yoon Pin Lim
ABSTRACT
Secreted proteins are an attractive minefield for cancer drug targets. An iTRAQ-based tandem mass spectrometry approach was employed to relatively quantify proteins in the secretomes of four isogenic breast cancer cell lines with increasing metastatic potential. CXCL3 was found to be upregulated in aggressive cancer cells. SiRNA and antibody neutralization studies supported a role of CXCL3 in metastatic processes. Meta-analysis of the mRNA level of CXCL3 in 1881 breast tumors supported a role of CXCL3 in clinical breast cancer. Our results support a functional role of CXCL3 in breast cancer metastasis and as a viable target for cancer therapy.
Sigma-Aldrich
MISSION® esiRNA, targeting human CXCL3
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Supelco
Monobasic Potassium Phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
USP
Monobasic potassium phosphate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%