Merck
Histone-like TAFs within the PCAF histone acetylase complex.
Cell (1998-07-23)
V V Ogryzko, T Kotani, X Zhang, R L Schiltz, T Howard, X J Yang, B H Howard, J Qin, Y Nakatani
ABSTRACT
PCAF histone acetylase plays a role in regulation of transcription, cell cycle progression, and differentiation. Here, we show that PCAF is found in a complex consisting of more than 20 distinct polypeptides. Strikingly, some polypeptides are identical to TBP-associated factors (TAFs), which are subunits of TFIID. Like TFIID, histone fold-containing factors are present within the PCAF complex. The histone H3- and H2B-like subunits within the PCAF complex are identical to those within TFIID, namely, hTAF(II)31 and hTAF(II)20/15, respectively. The PCAF complex has a novel histone H4-like subunit with similarity to hTAF(II)80 that interacts with the histone H3-like domain of hTAF(II)31. Moreover, the PCAF complex has a novel subunit with WD40 repeats having a similarity to hTAF(II)100.