Merck
  • Enhanced sensitivity detection of protein immobilization by fluorescent interference on oxidized silicon.
Enhanced sensitivity detection of protein immobilization by fluorescent interference on oxidized silicon.
Biosensors & bioelectronics (2003-11-19)
J-N Volle, G Chambon, A Sayah, C Reymond, N Fasel, M A M Gijs
ABSTRACT
We present a silicon chip-based approach for the enhanced sensitivity detection of surface-immobilized fluorescent molecules. Green fluorescent protein (GFP) is bound to the silicon substrate by a disuccinimidyl terephtalate-aminosilane immobilization procedure. The immobilized organic layers are characterized by surface analysis techniques, like ellipsometry, atomic force microscopy (AFM) and X-ray induced photoelectron spectroscopy. We obtain a 20-fold enhancement of the fluorescent signal, using constructive interference effects in a fused silica dielectric layer, deposited before immobilization onto the silicon. Our method opens perspectives to increase by an order of magnitude the fluorescent response of surface immobilized DNA- or protein-based layers for a variety of biosensor applications.
Sigma-Aldrich
N-Ethyldiisopropylamine solution, suitable for peptide synthesis, ~2 M in 1-methyl-2-pyrrolidinone