Merck
  • DEAD-box helicase DP103 defines metastatic potential of human breast cancers.
DEAD-box helicase DP103 defines metastatic potential of human breast cancers.
The Journal of clinical investigation (2014-08-02)
Eun Myoung Shin, Hui Sin Hay, Moon Hee Lee, Jen Nee Goh, Tuan Zea Tan, Yin Ping Sen, See Wee Lim, Einas M Yousef, Hooi Tin Ong, Aye Aye Thike, Xiangjun Kong, Zhengsheng Wu, Earnest Mendoz, Wei Sun, Manuel Salto-Tellez, Chwee Teck Lim, Peter E Lobie, Yoon Pin Lim, Celestial T Yap, Qi Zeng, Gautam Sethi, Martin B Lee, Patrick Tan, Boon Cher Goh, Lance D Miller, Jean Paul Thiery, Tao Zhu, Louis Gaboury, Puay Hoon Tan, Kam Man Hui, George Wai-Cheong Yip, Shigeki Miyamoto, Alan Prem Kumar, Vinay Tergaonkar
ABSTRACT
Despite advancement in breast cancer treatment, 30% of patients with early breast cancers experience relapse with distant metastasis. It is a challenge to identify patients at risk for relapse; therefore, the identification of markers and therapeutic targets for metastatic breast cancers is imperative. Here, we identified DP103 as a biomarker and metastasis-driving oncogene in human breast cancers and determined that DP103 elevates matrix metallopeptidase 9 (MMP9) levels, which are associated with metastasis and invasion through activation of NF-κB. In turn, NF-κB signaling positively activated DP103 expression. Furthermore, DP103 enhanced TGF-β-activated kinase-1 (TAK1) phosphorylation of NF-κB-activating IκB kinase 2 (IKK2), leading to increased NF-κB activity. Reduction of DP103 expression in invasive breast cancer cells reduced phosphorylation of IKK2, abrogated NF-κB-mediated MMP9 expression, and impeded metastasis in a murine xenograft model. In breast cancer patient tissues, elevated levels of DP103 correlated with enhanced MMP9, reduced overall survival, and reduced survival after relapse. Together, these data indicate that a positive DP103/NF-κB feedback loop promotes constitutive NF-κB activation in invasive breast cancers and activation of this pathway is linked to cancer progression and the acquisition of chemotherapy resistance. Furthermore, our results suggest that DP103 has potential as a therapeutic target for breast cancer treatment.
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder
Etoposide, European Pharmacopoeia (EP) Reference Standard
Etoposide for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(S)-(+)-Camptothecin, ≥90% (HPLC), powder