Merck
  • Enzymatic hydrolysis of nitriles by an engineered nitrile hydratase (papain Gln19Glu) in aqueous-organic media.
Enzymatic hydrolysis of nitriles by an engineered nitrile hydratase (papain Gln19Glu) in aqueous-organic media.
Biotechnology and bioengineering (2007-06-27)
Andrea Versari, Robert Ménard, Robert Lortie
ABSTRACT
A mutant of the cysteine protease papain, displaying nitrile hydratase and amidase activities, was expressed in Pichia pastoris and used for the hydrolysis of peptide nitriles in aqueous-organic media. The rate of hydrolysis of these nitriles is lowered by increasing acetone concentration. This is caused by an increase of the Michaelis constant, and a variation of Vmax proportional to the amount of water in the mixture. The hydrolysis of the amide is less affected by the increase in co-solvent, which results in lower accumulation of this intermediate product. With the peptide nitrile tested, high nitrile concentrations could be used to promote the production of the amide and prevent its hydrolysis to the acid by diminishing the relative rate of amide hydrolysis. A number of non-peptidyl nitriles were also tested as potential substrates but activity was detected for only one compound with structural resemblance to peptide nitriles.
Sigma-Aldrich
N-(Benzyloxycarbonyl)-2-aminoacetonitrile, 98%