- A Versatile Synthetic Extracellular Matrix Mimic via Thiol-Norbornene Photopolymerization.
A Versatile Synthetic Extracellular Matrix Mimic via Thiol-Norbornene Photopolymerization.
Advanced materials (Deerfield Beach, Fla.) (2009-12-28)
Benjamin D Fairbanks, Michael P Schwartz, Alexandra E Halevi, Charles R Nuttelman, Christopher N Bowman, Kristi S Anseth
PMID25377720
ABSTRACT
Step-growth, radically mediated thiol-norbornene photopolymerization is used to create versatile, stimuli-responsive poly(ethylene glycol)-co-peptide hydrogels The reaction is cytocompatible and allows for the encapsulation of human mesenchymal stem cells with a viability greater than 95%. Cellular spreading is dictated via three-dimensional biochemical photopatterning.