Merck
  • One-pot synthesis of 2-substituted indoles from 2-aminobenzyl phosphonium salts. A formal total synthesis of arcyriacyanin A.
One-pot synthesis of 2-substituted indoles from 2-aminobenzyl phosphonium salts. A formal total synthesis of arcyriacyanin A.
Organic letters (2008-06-25)
George A Kraus, Haitao Guo
ABSTRACT
The reaction of (2-aminobenzyl) triphenylphosphonium bromide with aromatic aldehydes or alpha,beta-unsaturated aldehydes under microwave-assisted conditions constitutes a new synthesis of 2-substituted indoles in high yields (81-97%) in a one-pot reaction. The adduct from indole-4-carboxaldehyde was an advanced intermediate in the synthesis of arcyriacyanin A.